KIMBLE® ULTRA-WARE® Microfiltration Assembly, DWK Life Sciences
À propos de cet article
This adapter assembly is designed to allow connection of a 47 mm or 90 mm filtration support base with a straight drip tip to a bottle or reservoir with a GL45 thread.
- Filtrate is collected directly into a media-type bottle when using a standard membrane filtration assembly
- All components are steam-autoclavable
ULTRA-WARE® microfiltration assembly with stainless steel support and GL 45 style bottle has a stainless steel support and is recommended for filtering viscous or proteinaceous solutions or to produce ultra-clean filtrate. This assembly has 80×80 304 stainless steel mesh, 0.0055" opening size, 19.4% open area, 0.007" wire diameter.
Certifications: Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.
Delivery information: Microfiltration assembly with stainless steel support supplied with funnel, clamp, 304 stainless steel support screen, PTFE gasket, glass support base, tubing adapter with o-ring, PBT cap, sealing ring and vacuum/pressure bottle.
2 Options Available Below
Product Details & Documents
This adapter assembly is designed to allow connection of a 47 mm or 90 mm filtration support base with a straight drip tip to a bottle or reservoir with a GL45 thread.
- Filtrate is collected directly into a media-type bottle when using a standard membrane filtration assembly
- All components are steam-autoclavable
ULTRA-WARE® microfiltration assembly with stainless steel support and GL 45 style bottle has a stainless steel support and is recommended for filtering viscous or proteinaceous solutions or to produce ultra-clean filtrate. This assembly has 80×80 304 stainless steel mesh, 0.0055" opening size, 19.4% open area, 0.007" wire diameter.
Certifications: Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.
Delivery information: Microfiltration assembly with stainless steel support supplied with funnel, clamp, 304 stainless steel support screen, PTFE gasket, glass support base, tubing adapter with o-ring, PBT cap, sealing ring and vacuum/pressure bottle.
Product Family Options
Informations sur le produit
- DésignationAvailabilityPrice
- Supplier Number: 179950-4532
- Microfiltration assembly with stainless steel support and GL45 style bottleLog in pour la disponibilitéLog in pour la disponibilitéEachEach - $1,358.00$1,358.00Prix promotionnel pour les nouveaux clientsEach$1,358.00Supplier Number: 953800-5347
Accessories
Informations sur le produit
- Catalog #AvailabilityPrice
- Filtration adpter assembly body, borosilicate glassFiltration adpter assembly body, borosilicate glassKT179951-3213Log in pour la disponibilitéEach - $109.65
Informations sur le produit
- Catalog #AvailabilityPrice
- PBT GL45 cap, 34 mm opening
Informations sur le produit
- Catalog #AvailabilityPrice
- Hose barb connector for filtration assembly, ¹/₄", 13-425Hose barb connector for filtration assembly, ¹/₄", 13-425KT736400-1413Log in pour la disponibilitéEach - $26.60
Informations sur le produit
- Catalog #AvailabilityPrice
- PTFE/silicone sealing ring, 42 mm
Informations sur le produit
- Catalog #AvailabilityPrice
- PTFE/silicone sealing ring, 29 mm
Informations sur le produit
- Catalog #AvailabilityPrice
- PTFE support screen gasket, 47 mm
Informations sur le produit
- Catalog #AvailabilityPrice
- Stainless steel support screen for 47 mm filtration assembly, 80×80 meshStainless steel support screen for 47 mm filtration assembly, 80×80 meshKT953810-0000Log in pour la disponibilitéEach - $525.74
Informations sur le produit
- Catalog #AvailabilityPrice
- Solvent bottle adapter, GL45