Mobile Stool Tables, Elliptical, AmTab | Avantor
Mobile Stool Tables, Elliptical, AmTab
   Options durables disponibles
Mobile Stool Tables, Elliptical, AmTab
   Options durables disponibles
Certificates
Mobile Stool Tables, Elliptical, AmTab
2 Options Available Below
Product Details & Documents
Mobile cafeteria tables are used every day throughout the world in multi-purpose environments. High-quality state-of-the-art tables are safe, durable, easy to operate, and a great long-term value. Tables are perfect for heavy-use and heavy-turn settings providing years of trusted service.
  • Affordable long-term value
  • 100% made in the USA
  • 15 year warranty
Custom high-speed gas cylinder stabilizer assist for long-lasting opening and closing functions and much more.
Certifications: UL certified, ANSI/BIFMA certified.
Specifications for Product Family
Spécifications
  • Catalog No:
  • 76178-670
  • 76178-668
  • Environmentally Preferable:
  • Safer Human and Environmental Health
  • Safer Human and Environmental Health
  • Environmental certification:
  • MAS GREEN
  • MAS GREEN
Spécifications
Product Family Options
Informations sur le produit
  • Désignation
    No. of Users
    W×D
    W×D×H
    Availability
    Price
  • Elliptical stool and bench combination
    12
    94×84"
    94×84"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $12,342.90
    $12,342.90
    Each
       $12,342.90
    La quantité minimale de commande est de 1
    Supplier Number: MSE1012
    Spécifications:
      Safer Human and Environmental Health
    More
    Specifications
    Cat. No.
    76178-670
    Environmentally Preferable
    Safer Human and Environmental Health
    Environmental certification
    MAS GREEN
      Safer Human and Environmental Health
  • Elliptical stool and bench combination
    10
    130×77"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $12,099.55
    $12,099.55
    Each
       $12,099.55
    La quantité minimale de commande est de 1
    Supplier Number: MSE610
    Spécifications:
      Safer Human and Environmental Health
    More
    Specifications
    Cat. No.
    76178-668
    Environmentally Preferable
    Safer Human and Environmental Health
    Environmental certification
    MAS GREEN
      Safer Human and Environmental Health