Mobile Stool Tables, 60'Ø, AmTab
Mobile Stool Tables, 60'Ø, AmTab
   Options durables disponibles
Mobile Stool Tables, 60'Ø, AmTab
   Options durables disponibles
Certificates
Mobile Stool Tables, 60'Ø, AmTab
2 Options Available Below
Product Details & Documents
Mobile cafeteria tables are used every day throughout the world in multi-purpose environments. High-quality state-of-the-art tables are safe, durable, easy to operate, perfect for heavy-use and heavy-turn settings providing years of trusted service.
  • Affordable long-term value
  • 100% made in the USA
  • 15 year warranty
Custom high-speed gas cylinder stabilizer assist for long-lasting opening and closing functions and much more.
Certifications: UL certified, ANSI/BIFMA certified.
Specifications for Product Family
Spécifications
  • Catalog No:
  • 76178-910
  • 76178-908
  • Environmentally Preferable:
  • Safer Human and Environmental Health
  • Safer Human and Environmental Health
  • Environmental certification:
  • MAS GREEN
  • MAS GREEN
Spécifications
Product Family Options
Informations sur le produit
  • Désignation
    No. of Users
    W×D
    Availability
    Price
  • Octagnal mobile stool and bench combination
    8
    85×85"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - 11 667,25 $
    11 667,25 $
    Each
       11 667,25 $
    Minimum order quantity is 1
    Supplier Number: MSOC608
    Spécifications:
      Safer Human and Environmental Health
    More
    Specifications
    Cat. No.
    76178-910
    Environmentally Preferable
    Safer Human and Environmental Health
    Environmental certification
    MAS GREEN
      Safer Human and Environmental Health
  • Round mobile stool and bench combination
    8
    85×85"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - 11 667,25 $
    11 667,25 $
    Each
       11 667,25 $
    Minimum order quantity is 1
    Supplier Number: MSR608
    Spécifications:
      Safer Human and Environmental Health
    More
    Specifications
    Cat. No.
    76178-908
    Environmentally Preferable
    Safer Human and Environmental Health
    Environmental certification
    MAS GREEN
      Safer Human and Environmental Health