Multivapor™ Parallel Evaporation Systems, Buchi
À propos de cet article
Evaporate up to 12 samples simultaneously by efficient vacuum vortex technology.
- Fast evaporation of multiple samples
- Easy operation
- Integration with rotavapor
- Full visibility of samples
Speed up your sample evaporation for multiple samples. With its ease of use, the Multivapor™ is tailored to maximize efficiency. A wide range of available adapters allow neat integration of your glassware into the daily routine.
2 Options Available Below
Product Details & Documents
Evaporate up to 12 samples simultaneously by efficient vacuum vortex technology.
- Fast evaporation of multiple samples
- Easy operation
- Integration with rotavapor
- Full visibility of samples
Speed up your sample evaporation for multiple samples. With its ease of use, the Multivapor™ is tailored to maximize efficiency. A wide range of available adapters allow neat integration of your glassware into the daily routine.
Product Family Options
Informations sur le produit
- DésignationCondenser TypeNo. of PositionsAvailabilityPrice
- Supplier Number: MP12101C22
- Supplier Number: MP12101S20
- Uses Rotavapor®12Discontinued-Supplier Number: -