Face Masks Dispenser, AK LTD.
Catalog # 32799-010
Supplier: AK
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
Specifications
- Dispenser type:Floor-standing/wall-mountable
- Material:PETG
- No. of compartments:1
- PPE type:Face masks
- Color:Clear
- Description:Face mask dispenser, PETG
- Environmentally Preferable:
- Thickness:³/₁₆"
- Cat. no.:32799-010
- W×D×H:229×134×204 mm
- Supplier No.:AK-266
Specifications
About this item
Face masks can be easily and carefully stored in this dispenser.
- Wall-mounted for free standing
- Handy three pre-pierced holes
- Clear design allowing for simpler use
- Hinged lid allows for easy access
The base opening allows for one at a time interval.