ContactUsLinkComponent
LopHene® Ⅱ Concentrated Low-pH Disinfectant, Decon Labs
LopHene® Ⅱ Concentrated Low-pH Disinfectant, Decon Labs
Catalog # 76214-018
Supplier:  Decon Labs
undefined
LopHene® Ⅱ Concentrated Low-pH Disinfectant, Decon Labs
Catalog # 76214-018
Supplier:  Decon Labs
Supplier Number:  8811
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    LopHene Ⅱ Concentrated Phenolic Disinfectant
  • Size:
    3.8 l (1.0 gal.)
  • Cat. no.:
    76214-018
  • Packaging:
    Poly bottle
Specifications
About this item
LopHene® Ⅱ concentrated phenolic germicide for rotational disinfection.
  • High soil-load and hard water tolerance
  • Diluted one ounce per gallon of water (1:128)
It can be used on hard surfaces in labs, production areas and on process equipment or anywhere thorough disinfection efficacy is required. LopHene II is a biological decontamination product and disinfection. It is effective against a wide range of bacteria, viruses and fungi, including TB and HIV-1.
Certifications: Conforms to OSHA’s blood borne Pathogens Standard. EPA registered.
Ordering information:
Available in convenient packaging configurations. Custom solutions or packaging configuration may be available. Please note that pure ethanol is regulated by the US ATF/TTB and depending on the taxable status of your institution, pure ethanol products are taxed according to the government schedule of taxes detailed in 27 CFR Chapter 1 section 27.41. To purchase tax-free, you must have an Industrial Alcohol User Permit from the AFT/TTB, specifically marked for “the withdraw and use of alcohol free of tax”. Tax-free pure ethanol is intended for industrial use only.