Fluorescent Clamp Base Task Light, Dazor Lighting Technology
Fluorescent Clamp Base Task Light, 33", Cool White, Full Spectrum or Daylight, 45 W
Catalog # 76246-718
Supplier: DAZOR LIGHTING TECHNOLOGY TE
Fournisseur #: 124C3-BLB
Fluorescent Clamp Base Task Light, Dazor Lighting Technology
Catalog # 76246-718
Supplier: DAZOR LIGHTING TECHNOLOGY TE
Jump to:
Product Details & Documents
This fluorescent light provides abundant light over a large work area with the benefits of a stylish and energy-efficient design.
- Provides abundant, even and shadow-free light over a broad surface
- All metal construction for rugged, heavy use environments
- Sturdy mount can be clamped or bolted to a horizontal surface
- Fully enclosed counterbalance spring for dust-free operation and low maintenance
- Durable powder-coat finish
- Five-year warranty
Features floating adjustable arm for smooth and easy positioning of the lamp to any position for a total reach of 33". State-of-the-art electronic ballast provides the highest energy-efficiency, instant on/off control of the bulbs and the elimination of visible bulb strobing or flickering.
Certifications: UL, cUL, CE.
Spécifications
- Désignation:Fluorescent Clamp Base Task Light, 33"
- Puissance:45 W
- Bulb Type:Cool White, Full Spectrum or Daylight