Envelope Clamp Gaskets, Garlock Hygienic Technologies
Envelope Clamp Gaskets, Garlock Hygienic Technologies
Supplier:   Rubber Fab
Envelope Clamp Gaskets, Garlock Hygienic Technologies
Supplier:  Rubber Fab
Certificates
Envelope Clamp Gaskets, Garlock Hygienic Technologies
2 Options Available Below
Product Details & Documents
These gaskets are used in processing pipeline systems for the food, dairy, beverage, pharmaceutical and bio tech industries.
 : Meets criteria of the FDA, and accepted by the USDA, and USP Class VI certifications.
Product Family Options
Informations sur le produit
  • Désignation
    Matière
    For Use With
    Availability
    Price
  • Mini envelope clamp gasket
    Teflon® fluoropolymer
    0.75"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $66.54
    $66.54
    Each
       $66.54
    Supplier Number: A42MPGR-TF-075-E
  • Envelope clamp gasket
    Teflon® fluoropolymer
    4"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $154.97
    $154.97
    Each
       $154.97
    Supplier Number: A40MPGR-TF-400-E