Dispensing Tips, EFD
Dispensing Tips, EFD
Dispensing Tips, EFD
Certificates
Dispensing Tips, EFD
2 Options Available Below
Product Details & Documents
EFD produces the highest quality dispensing tips engineered hub flats for easy twist on, twist off. 360° SafetyLok™ thread ensures safe, positive attachment to syringe barrel.
  • Tapered Smooth flow for application of medium
  • Flexible Polypropylene shafts reach into hard-to-access areas
  • Free of flash, burrs and contaminants
  • Package labels include lot numbers for process control and traceability
For applications in chamfered, ESD-safe, PTFE-coated and PTFE-lined, microdot tips and oval tips.
Product Family Options
Precision Dispensing Tip
Informations sur le produit
  • Désignation
    Jauge
    Longueur
    Ø ext.
    Ø int.
    Color
    Availability
    Price
  • Precision stainless steel dispensing tip, with 45° bend
    15
    12.7 mm (0.50")
    1.65 mm (0.065")
    1.36 mm (0.053")
    Amber
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $162.18
    $162.18
    Each
       $162.18
    Supplier Number: 7018069
  • Precision stainless steel dispensing tip, with 45° bend
    20
    12.7 mm (0.50")
    0.91 mm (0.036")
    0.61 mm (0.024")
    Pink
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $257.75
    $257.75
    Each
       $257.75
    Supplier Number: 7018179