Proto® Socket Bit, Grainger
Proto® Socket Bit, Six Points, Alloy steel, 1⅞", 1/4" Hex, Full polished chrome, 5/64"
Catalog # 76253-508
Supplier: GRAINGER
Supplier #: 426H51
Jump to:
Product Details & Documents
The Proto® socket bit has a hex opening that allows the bit holder to take the torque load.
- Polished chrome finish
- Insert length: 1"
Made from a heat-treated alloy for wear resistance which minimizes distortion for a long life.
Specifications
- Désignation:Proto® Socket Bit, Six Points
- Longueur:1⅞"
- Matière:Alloy steel
- Tip Size:5/64"
- Finish:Full polished chrome
- Drive Size:1/4" Hex