Scribers, TESTEQUITY LLC TE | Avantor
Scribers, TESTEQUITY LLC TE
8.5", Scribe, straight angled 90° with knurled handle
Catalog # 76244-434
Supplier:   TESTEQUITY LLC TE
Fournisseur #:  76-1510
Scribers, TESTEQUITY LLC TE
Catalog # 76244-434
Supplier:  TESTEQUITY LLC TE
Scribers, TESTEQUITY LLC TE
8.5", Scribe, straight angled 90° with knurled handle
Price
$14.07
Log in pour la disponibilité
Each
Product Details & Documents
Spécifications
  • Longueur:
    8.5"
  • Includes:
    Scribe, straight angled 90° with knurled handle