Solid Carbide Square End Mill, Spiral Flute, Sid Tool
Square End Mill, 3 Flute, Single End, Solid Carbide, 3/16", ¾", Uncoated, ³⁄₁₆"
76251-098
: 87810487
Solid carbide provides better rigidity than high speed steel. It is extremely heat resistant and used for high speed applications on cast iron, nonferrous materials, plastics and other tough-to-machine materials.
- Uncoated tools do not feature supportive treatments on the cutting edge
- Used at reduced speeds for general applications on nonferrous metals
- Uncoated, spiral flute, 40° helix
- Centercutting with right hand cut
- Right hand flute, long length
Center cutting tools have one or more cutting edges at the tip and are used in a variety of applications including plunging, drilling or ramping.
- Désignation:Square End Mill, 3 Flute, Single End
- Longueur:¾"
- Matière:Solid Carbide
- Finish:Uncoated
- Shank Diameter:³⁄₁₆"
- Diameter:3/16"