ProGuard™ Screwdrivers, Global
11", 13½", 3/32"
  76250-186
 :  3705
ProGuard™ Screwdrivers, Global
  76250-186
ProGuard™ Screwdrivers, Global
11", 13½", 3/32"
$55.12
Each
It is an extra long length ball end screwdriver.
  • Tip Size:
    3/32"
  • Handle Length:
    11"
  • Overall Length:
    13½"