Plug and Pin Gage, Sid Tool
Plug and Pin Gage, 0.08"
Catalog # 76251-568
Supplier: SID TOOL CO INC TE
Supplier #: 84499193
Jump to:
Product Details & Documents
Plug and Pin Gage, Class ZZ Plus, with tolerence of 0.0002" and black oxide finish.
Specifications
- Ø:0.08"
- Désignation:Plug and Pin Gage