Reusable Altessa Grid Pull-Over Cleanroom Hoods, Uniform Technology
Cleanroom hoods are manufactured from 99% polyester/1% carbon fabric.
- Pull-over, open face design
- Vertical and horizontal snaps on back
- Snaps at temples
- Split skirt
Suitable for ISO Class 5 or higher cleanrooms.
- Catalog No:
- 76268-140
- 76268-138
- 76515-812
- 76515-810
- 76515-808
- 76515-806
- 76515-804
- 76515-802
- 76515-800
- 76515-798
- 76515-796
- 76268-142
- 76515-794
- 76515-792
- Garment application:
- Cleanroom
- Cleanroom
- Cleanroom
- Cleanroom
- Cleanroom
- Cleanroom
- Cleanroom
- Cleanroom
- Cleanroom
- Cleanroom
- Cleanroom
- Cleanroom
- Cleanroom
- Cleanroom
- Fabric:
- 100% polyester Taffeta
- 100% polyester Taffeta
- 100% polyester Taffeta
- 100% polyester Taffeta
- 100% polyester Taffeta
- 100% polyester Taffeta
- 100% polyester Taffeta
- 100% polyester Taffeta
- 100% polyester Taffeta
- 100% polyester Taffeta
- 100% polyester Taffeta
- 100% polyester Taffeta
- 100% polyester Taffeta
- 100% polyester Taffeta
Frequently Bought Together