Kelvin Probes, TESTEQUITY LLC TE
Catalog # 76251-530
Supplier: TESTEQUITY LLC TE
Specifications
- Description:Kelvin Probe Set with Double Banana Plug Leads
- Length:48"
- Cat. no.:76251-530
Specifications
About this item
Probes for 4-wire and 2-wire measurements.
- Patented design for high precision rapid testing
- Gold-plated for accuracy
- Easily move from one test point to another for rapid readings
- For use with 4-wire Kelvin meters