Shelves, Production Basics
Supplier: Production Basics
Certificates
About this item
The universal versatile metal workstation shelf can be installed in three ways. Use as a traditional flat shelf, extend it behind the workbench frame or tilt it up to 30 degrees.
- Finished with rounded corners and protective T-mold edge
Laminate under surface shelf mounts to the frame under the worksurface and provides additional storage space.
The heavy-duty laminate shelf for applications requiring up to 175 lbs. of support.
Frequently Bought Together