ESD Floor Mat Grounding Kit, Transforming Technologies
Catalogue # 76250-302
Fournisseur: Transforming Technologies
Spécifications
- Color:Black
- Description:Floor Mat Ground Kit
- Cat. no.:76250-302
Spécifications
À propos de cet article
This ground Kit comes with a molded low profile ground cord with mushroom cap and a 3/8 (10 mm) snap ensures secure connection to floor mats.
Certifications: Meets or exceeds requirements of ANSI ESD-S20.20 and ANSI/ESD S-6.1.