Forceps - Serrated Tips
Forceps with bent serrated tips, 12" (304.8 mm)
78620-536
New Product
: 18493
General forceps with serrated tips.
Stainless steel finish provides excellent corrosion resistance
Ideal for bending, separating and removing small items
Long, straight tip is helpful when working in confined spaces
- Désignation:Forceps with bent serrated tips
- Longueur:12" (304.8 mm)