Extension Cords, Mountz
Extension cord, Black, 3 m (10 ft.)
76251-270
: 144180
Can be combined with existing power cord.
Works with A, BL, CL, EF, ESD, K or SS Series Drivers.
- Color:Black
- Désignation:Extension cord
- Longueur:3 m (10 ft.)
Can be combined with existing power cord.
Works with A, BL, CL, EF, ESD, K or SS Series Drivers.