Envelope Gaskets, Garlock Hygienic Technologies
 :   Rubber Fab
Envelope Gaskets, Garlock Hygienic Technologies
Envelope Gaskets, Garlock Hygienic Technologies
A PTFE envelope gasket has the chemical resistance of PTFE and the elastomeric characteristics of FKM Fluoroelastomer.
 : Meets FDA and USP Class VI standards.
Product Family Options
  • Désignation
    Matière
    Taille
  • Envelope gasket
    PTFE
    1.0"
    Each
    Each - $14.93
    $14.93
    Each
       $14.93
     : A40MPGR-100-V
  • Envelope gasket
    PTFE
    1.5"
    Each
    Each - $11.25
    $11.25
    Each
       $11.25
     : A40MPGR-150-V
  • Envelope gasket
    PTFE
    2.0"
    Each
    Each - $13.33
    $13.33
    Each
       $13.33
     : A40MPGR-200-V