ContactUsLinkComponent
Electronic Probes, TESTEQUITY LLC TE
Catalog # 76242-778
Supplier:  TESTEQUITY LLC TE
Electronic Probes, TESTEQUITY LLC TE
Catalog # 76242-778
Supplier:  TESTEQUITY LLC TE
Supplier Number:  6341
Specifications
  • Description:
    Precision "Pogo-Pin" Electronic Probe Kit
  • Cat. no.:
    76242-778
Specifications
About this item
Pogo pin spring-loaded tips. Plugs fit most Fluke, Tektronix, Hewlett-Packard and Wavetek meters.
Delivery: Kit contains a pair (black/red) of precision electronic probes, and five sets of 0.040" diameter. Replacement tips. Also included are (2 each): 1" long stainless steel tips, 3" long stainless steel tips, gold-plated plunger tips with sharp point, gold-plated 3-point plunger tips, gold-plated 4-point plunger tips and plastic case. Rated for 1000 V, 3 A maximum.