Adjustable Wrist Straps with Dual Wire Coil Cords, Botron
Fournisseur: BOTRON COMPANY INC.
Certificats
À propos de cet article
High quality adjustable dual anti static wrist strap sets, they provide the performance and reliability of a metal band with the comfort of cloth. The blue knitted material has a soft outer area and a 5/8" conductive area on the inside.
Frequently Bought Together