Document Holders, Statfree, Protektive Pak
Document Holders with Extra Pocket, 6 mil 1.7 lbs., 10×12"
Catalog # 76239-988
Supplier: Protektive Pak
Fournisseur #: 47122
Jump to:
Product Details & Documents
This holder used in static sensitive testing and production environments.
- Imprinted with ESD protective symbol
- Holds work orders and drawings
Low charging and dissipative, provides path-to-ground.
Spécifications
- Désignation:Document Holders with Extra Pocket, 6 mil 1.7 lbs.
- Dimensions:10×12"