Casters, Zoro
Caster wheel, 5", brown, 450 lbs.
Catalog # 76252-108
Supplier: ZORO - CUSTOMER SERVICE TE
Supplier #: G4824687
Jump to:
Product Details & Documents
Specifications
- Désignation:Caster wheel, 5", brown
- Load Capacity:450 lbs.