Collapsible Bulk Box, Quantum Storage Systems
1800 lbs./l, Collaspsible Bulk Box, 48×45×34" (E×terior); 441/2×411/2×281/5" (Interior)
Catalog # 76248-966
Supplier: QUANTUM STORAGE SYSTEMS
Fournisseur #: QBB-4845-34
Jump to:
Product Details & Documents
The collapsible bulk box is your one-stop transportation container. Manufactured from high-density materials, this bulk box protects all goods packed within during transit or in storage. When not in use, the container can be collapsed to a fraction of its size providing more space and compact transportation.
- Ergonomic Latches for opening and closing
Spécifications
- Capacité:1800 lbs./l
- Désignation:Collaspsible Bulk Box
- Dimensions:48×45×34" (E×terior); 441/2×411/2×281/5" (Interior)