CFP Boxes, Conductive Containers
CFP Box holds 3 Boxes
Catalog # 76249-002
Supplier: CONDUCTIVE CONTAINERS INC. TE
Fournisseur #: 08M-9084-CFP
Jump to:
Product Details & Documents
Conductive fluted plastic (CFP) containers with winged partitions and two strips of hooks and velcro on bottoms.
- Thickness of 3 mm
Spécifications
- Désignation:CFP Box holds 3 Boxes