Storage Case, Midwest Container Systems | Avantor
Storage Case, Midwest Container Systems
Storage Case, 85/8×51/8×15/8"
Catalog # 76245-324
Fournisseur #:  5705
Storage Case, Midwest Container Systems
Catalog # 76245-324
Storage Case, Midwest Container Systems
Storage Case, 85/8×51/8×15/8"
Price
40,51 $
Log in pour la disponibilité
Each
Product Details & Documents
Plastic storage case include adjustable dividers for customized storage of small parts and supplies.
  • Made of clear industrial-grade polymers for durability and visibility of contents
  • Molded-in "feet" allow cases to sit upright
A unique grid design in the lid prevents items from spilling into other compartments.
Spécifications
  • Désignation:
    Storage Case
  • Dimensions:
    85/8×51/8×15/8"