Standard Hand Tap, Sid Tool
Straight Flute Standard Hand Tap, with M18×1.00 thread, 4¹⁄₃₂", 1¹³⁄₁₆"
Catalog # 76249-004
Supplier: SID TOOL CO INC TE
Fournisseur #: 68762897
Jump to:
Product Details & Documents
Straight four flute standard hand tap, with bottoming chamfers are used for threading blind holes. They have 1 to 2 threads.
- High speed steel (HSS) provides good wear-resistance
- Used in general purpose applications for both ferrous and nonferrous materials
- Bright provides a smooth, polished finish on the tool
It increases chip flow in softer materials such as aluminum, wood and plastic.
Spécifications
- Désignation:Straight Flute Standard Hand Tap, with M18×1.00 thread
- Longueur:4¹⁄₃₂"
- Thread Length:1¹³⁄₁₆"