Adapters, Digi-Key
Adapter Connector, 0.126" (3.20 mm) I-Ø, 0.327" (8.30 mm) O-Ø
Catalog # 76250-608
Supplier: DIGI-KEY CORPORATION
Supplier #: BPJJ02AUX
Jump to:
Product Details & Documents
Adapter connector Phono (RCA) jack to Phono (RCA) jack panel mount.
- Brass body with gold finish
- Nickel plated contact material
- Red insulation
- Unshielded
Female to female connector.
: Includes 1 connector, 1 nut and 1 washer.
Specifications
- Désignation:Adapter Connector, 0.126" (3.20 mm) I-Ø, 0.327" (8.30 mm) O-Ø