ADC® Pros Combo III Pocket Aneroid Kits
Fournisseur: AMERICAN DIAGNOSTIC CORP CP
Certificats
À propos de cet article
Ideal for Nurses, PA's, and EMT's.
- ADC®'s best pocket aneroid and most popular scope
- Includes extra-large color matched carry case
- Meets or exceeds ANSI/AAMI/ISO 81060-1
- Assembled, inspected, and packaged in the USA
For the discriminating professional who prefers a more traditional design, this combo featuring the 700 series chrome-plated aneroid and Adscope® 603, is perfect. The pocket aneroid has chrome plated hardware, and a luminescent dial with a manometer backed with a lifetime calibration warranty. The nylon cuff is marked with ADC®'s size guide to prevent miscuffing. The Adscope® 603 stethoscope utilizes AFD technology for the best acoustic response, and stainless steel binaurals fixed at 15O for greater comfort. These two items are color matched and supplied with an extra-large, dual pocket carrying case.
Frequently Bought Together