Tryptic Soy Broth, USP, 15 ml Vial with Needle-Port Septum
Catalogue # CA76006-258
Fournisseur: HARDY DIAGNOSTICS CA
Numéro CAS:
Spécifications
- Pack type:Vial
- Description:Tryptic soy broth, USP
- Size:10/pack
- Cat. no.:CA76006-258
- Fill Volume:15 ml
- Application:Cosmetics, Pharmaceutical manufacturing
Spécifications
À propos de cet article
Tryptic soy broth is recommended for use as a general purpose medium for the isolation and cultivation of a wide variety of bacteria and fungi. The U.S. Pharmacopeia national formulary (USP) describes its use for sterility testing.
- Surface sampling
- Air sampling
- Sterility tests
- Quality culture media tested for performance, pH, and sterility
- Kits are designed to help you follow USP <797> guidelines
- Kits supply what is needed to test the proficiency and aseptic technique of technicians or pharmacists in a self-contained ready-to-use format
- Kits are available for low, medium, and high complexity procedures
Hardy Diagnostics offers all the products you need to easily assess the risk of microbial contamination of your CSPs (Compounded Sterile Preparations) according to USP Chapter <797>.
Certifications: Easily assess the risk of microbial contamination of CSPs (Compounded Sterile Preparations) according to USP Chapter 797.