Ao Absorbance Microplate Reader, Azure Biosystems
About this item
96 well plate absorbance microplate reader.
- Offers a simple user interface
- Absorbance-based reading offers maximum sensitivity
- Best read speeds for kinetic assays
- High flexibility
- Lower cost compared to fluorescent and luminescent-based readers
Its top-reading design is ideal for solution-based assays such as ELISAs, DNA and protein quantification, and enzyme kinetics.
Stand-alone, top absorbance reader. Large, easy-to-use 7" touch screen interface. Pre-set and custom protocols. Single and dual wavelength detection modes. 8 position filter wheel: 5 standard filters (405, 450, 492, 595, and 630) plus 3 options from 340 to 750 nm. Built-in shaker with speed selection. Motorized door for easy sample loading.
4 Options Available Below
Product Details & Documents
96 well plate absorbance microplate reader.
- Offers a simple user interface
- Absorbance-based reading offers maximum sensitivity
- Best read speeds for kinetic assays
- High flexibility
- Lower cost compared to fluorescent and luminescent-based readers
Its top-reading design is ideal for solution-based assays such as ELISAs, DNA and protein quantification, and enzyme kinetics.
Stand-alone, top absorbance reader. Large, easy-to-use 7" touch screen interface. Pre-set and custom protocols. Single and dual wavelength detection modes. 8 position filter wheel: 5 standard filters (405, 450, 492, 595, and 630) plus 3 options from 340 to 750 nm. Built-in shaker with speed selection. Motorized door for easy sample loading.
Product Family Options
Filters
Product Information
- DésignationIncludesAvailabilityPrice
- Ao 340 nm FilterDiscontinued-Supplier Number: AC3100Specifications:
More
Specifications
Cat. No.76291-986 - Ao 380 nm FilterDiscontinued-Supplier Number: AC3101Specifications:
More
Specifications
Cat. No.76291-988 - Ao 405 nm FilterDiscontinued-Supplier Number: AC3102Specifications:
More
Specifications
Cat. No.76291-990 - Supplier Number: AC3103Specifications:
More
Specifications
Cat. No.76291-992 - Ao 450 nm FilterDiscontinued-Supplier Number: AC3104Specifications:
More
Specifications
Cat. No.76291-994 - Supplier Number: AC3115Specifications:
More
Specifications
Cat. No.76291-996 - Supplier Number: AC3116Specifications:
More
Specifications
Cat. No.76291-998 - Ao 492 nm FilterDiscontinued-Supplier Number: AC3105Specifications:
More
Specifications
Cat. No.76292-000 - Ao 510 nm FilterDiscontinued-Supplier Number: AC3117Specifications:
More
Specifications
Cat. No.76292-002 - Ao 520 nm FilterDiscontinued-Supplier Number: AC3118Specifications:
More
Specifications
Cat. No.76292-040 - Ao 525 nm FilterDiscontinued-Supplier Number: AC3119Specifications:
More
Specifications
Cat. No.76292-042 - Ao 532 nm FilterDiscontinued-Supplier Number: AC3120Specifications:
More
Specifications
Cat. No.76292-044 - Ao 540 nm FilterDiscontinued-Supplier Number: AC3106Specifications:
More
Specifications
Cat. No.76292-046 - Ao 545 nm FilterDiscontinued-Supplier Number: AC3121Specifications:
More
Specifications
Cat. No.76292-004 - Ao 560 nm FilterDiscontinued-Supplier Number: AC3122Specifications:
More
Specifications
Cat. No.76292-006 - Ao 562 nm FilterDiscontinued-Supplier Number: AC3123Specifications:
More
Specifications
Cat. No.76292-008 - Ao 570 nm FilterDiscontinued-Supplier Number: AC3107Specifications:
More
Specifications
Cat. No.76292-010 - Ao 578 nm FilterDiscontinued-Supplier Number: AC3108Specifications:
More
Specifications
Cat. No.76292-012 - Ao 590 nm FilterDiscontinued-Supplier Number: AC3109Specifications:
More
Specifications
Cat. No.76292-014 - Ao 595 nm FilterDiscontinued-Supplier Number: AC3110Specifications:
More
Specifications
Cat. No.76292-016 - Ao 600 nm FilterDiscontinued-Supplier Number: AC3124Specifications:
More
Specifications
Cat. No.76292-018 - Ao 620 nm FilterDiscontinued-Supplier Number: AC3125Specifications:
More
Specifications
Cat. No.76292-020 - Ao 630 nm FilterDiscontinued-Supplier Number: AC3111Specifications:
More
Specifications
Cat. No.76292-022 - Ao 646 nm FilterDiscontinued-Supplier Number: AC3126Specifications:
More
Specifications
Cat. No.76292-024 - Ao 650 nm FilterDiscontinued-Supplier Number: AC3112Specifications:
More
Specifications
Cat. No.76292-026 - Ao 663 nm FilterDiscontinued-Supplier Number: AC3127Specifications:
More
Specifications
Cat. No.76292-028 - Ao 690 nm FilterDiscontinued-Supplier Number: AC3113Specifications:
More
Specifications
Cat. No.76292-030 - Ao 700 nm FilterDiscontinued-Supplier Number: AC3128Specifications:
More
Specifications
Cat. No.76292-032 - Ao 750 nm FilterDiscontinued-Supplier Number: AC3114Specifications:
More
Specifications
Cat. No.76292-034
Printing
Product Information
- DésignationIncludesAvailabilityPrice
- Ao USB Thermal PrinterUSB Thermal Printer for Azure Ao Absorbance Microplate ReaderLog in for availabilityEach$382.38Supplier Number: AC3200Specifications:
More
Specifications
Cat. No.76292-036 - Ao Thermal PaperThermal Paper for Azure Ao Absorbance Microplate Reader USB Thermal PrinterDiscontinued-Supplier Number: AC3201Specifications:
More
Specifications
Cat. No.76292-038
Services
Product Information
- Catalog #DescriptionAvailabilityPrice