ContactUsLinkComponent
Jacketed Beakers/Baths, Low Form, Flat Bottom, Chemglass
Catalogue # 89028-334
Fournisseur:  Chemglass
Fournisseur #: CG-1107-11
Small Business Enterprise
undefined
Jacketed Beakers/Baths, Low Form, Flat Bottom, Chemglass
Catalogue # 89028-334
Fournisseur:  Chemglass
Supplier Number:  CG-1107-11
Fournisseur #: CG-1107-11
Small Business Enterprise
S'identifier pour la disponibilité
Select an Option
75/95 mm
Select an Option
Jacketed Beaker, Low Form, Flat Bottom
Select an Option
150/190 mm
Select an Option
89028-334
Select an Option
CG-1107-11
À propos de cet article
Jacketed beakers/baths with a larger opening at the top of the jacket and a shorter overall height.
  • Width to Height ratio on all sizes is 2:1
  • Flat bottom on both the inner and outer vessel
  • Designed for use in temperature controlled chemical and biological reactions
Hose connections have an O.D. of 10 mm at the largest serration.
Frequently Bought Together