ContactUsLinkComponent
Liquid Level Indicators, J.T.Baker®, Avantor
Catalogue # CAJT9803-03
MOC:  Non réglementé
Liquid Level Indicators, J.T.Baker®, Avantor
Catalogue # CAJT9803-03
Supplier Number:  BAKR9803-03
MOC:  Non réglementé
Spécifications
  • Description:
    Cycle-Tainer™ LCD liquid level volume display, for 52 L model S system
  • Cat. no.:
    CAJT9803-03
Spécifications
À propos de cet article
These level indicators provide the digital readout of the approximate volume of solvent remaining in the CYCLE-TAINER system.
  • Signal can also be used to drive switching circuits