ContactUsLinkComponent
Quartz Griffin Beakers, Chemglass
Quartz Griffin Beakers, Chemglass
Catalog # 89028-378
Supplier:  Chemglass
Supplier #: CG-1110-04
Small Business Enterprise
undefined
Quartz Griffin Beakers, Chemglass
Catalog # 89028-378
Supplier:  Chemglass
Supplier Number:  CG-1110-01
Supplier #: CG-1110-04
Small Business Enterprise
Log in for availability
Select an Option
80 mm
Select an Option
400 ml
Select an Option
Quartz Beaker
Select an Option
105 mm
Select an Option
89028-378
Select an Option
CG-1110-04
About this item
Low form beakers made of quartz.
  • Non-graduated with pouring spout
  • Beaker can be used at temperatures up to 1000 °C
Frequently Bought Together