Caps for 13-425 Screw-Thread Vials
Caps for 13-425 Screw-Thread Vials
Caps for 13-425 Screw-Thread Vials
Supplier:  Thermo Scientific
Certificates
Caps for 13-425 Screw-Thread Vials
3 Options Available Below
Product Details & Documents
Screw thread caps feature a 13-425 thread finish and are available in an assortment of styles to match your HPLC, GC or IC application needs.
  • Polypropylene caps are chemically inert and suitable for most chromatography applications
  • Phenolic resin caps perform well at high temperatures and are compatible with exposure to corrosives
  • Caps are bonded septa resist dislodging during injection when using large diameter blunt needles
Convenient pre-assembled caps and septa minimize contamination from handling.
Product Family Options
Product Information
  • Désignation
    Septum
    Taille du bouchon
    Cap Color
    Cap Type
    Availability
    Price
  • Open-top screw cap
    None
    13-425
    Black
    PP cap
    Log in for availability
    Log in for availability
    Log in for availability
    Pack of 100
    Pack of 100 - $48.87
    $48.87
    New Customer Promotional Price
    Log in for availability
    Case of 1000
    Case of 1000 - $488.68
    $488.68
    New Customer Promotional Price
    Pack of 100
       $48.87
    Case of 1000
       $488.68
    Supplier Number: C4015-1A
  • Open-top screw cap
    None
    13-425
    Black
    Phenolic cap
    Log in for availability
    Log in for availability
    Case of 1,000
    Case of 1,000 - $427.45
    $427.45
    New Customer Promotional Price
    Case of 1,000
       $427.45
    Supplier Number: C4015-66
  • Open-top screw cap
    Red PTFE/white silicone
    13-425
    Black
    Phenolic cap
    Log in for availability
    Log in for availability
    Pack of 100
    Pack of 100 - $231.56
    $231.56
    New Customer Promotional Price
    Pack of 100
       $231.56
    Supplier Number: C4015-66A
  • Yellow
    Discontinued
    Supplier Number: -
  • Black
    Discontinued
    Supplier Number: -