Glas-Col® Series O Heating Mantles, for Cylindrical Vessels with Bottom Drain Openings, Chemglass
Supplier: Chemglass
Certificates
About this item
These Glas-Col® Series O mantles are similar to the CG-10007 series. However this mantle is slit lengthwise to a 1.5" hole in the bottom center with lacing closure to accommodate a cylindrical reaction vessel (CG-1924) with a bottom drain.
- Mantle is glass fabric
- 115 volts
- Supplied with a separable 4', 2-wire extension cord with locking connector for each circuit
The 10,000 ml size has two circuits at the listed wattage.
Frequently Bought Together