VWR® Moisture Analyzers
À propos de cet article
The VWR® series Moisture Analyzers offer precise and reliable results for moisture content analysis.
- Automatic opening and closing of drying chamber lid
- Large, 5" color capacitive touch screen
- Moisture content analysis
- Dry mass determination
- Hands-free operation with IR proximity sensors
- Three-year warranty
The VWR® Moisture Analyzers can be utilized for a range of applications. Choose between an IR Emitter (max temperature 160 °C) or Halogen (max temperature 250 °C) heating module. For accurate moisture content analysis, the VWR® moisture analyzer features 4 drying profiles (standard, fast, step mild) and 4 finish modes (manual, automatic, time-defined, user-defined) to allow for timely, repeatable results.
The large, color capacitive touch screen allows for easy navigation of the VWR® moisture analyzers menus. The IR proximity sensors allow for hands free operation, easily program the sensors to perform actions such as opening/closing drying chamber lid, zeroing, taring, or starting an experiment.
Store up to 5000 products, 200 drying modes, 5000 drying reports, and 100 users within the VWR® Moisture Analyzers databases.
Certifications: C-UL(US) and CE.
Delivery information: Includes moisture analyzer and aluminum pan.
3 Options Available Below
Product Details & Documents
The VWR® series Moisture Analyzers offer precise and reliable results for moisture content analysis.
- Automatic opening and closing of drying chamber lid
- Large, 5" color capacitive touch screen
- Moisture content analysis
- Dry mass determination
- Hands-free operation with IR proximity sensors
- Three-year warranty
The VWR® Moisture Analyzers can be utilized for a range of applications. Choose between an IR Emitter (max temperature 160 °C) or Halogen (max temperature 250 °C) heating module. For accurate moisture content analysis, the VWR® moisture analyzer features 4 drying profiles (standard, fast, step mild) and 4 finish modes (manual, automatic, time-defined, user-defined) to allow for timely, repeatable results.
The large, color capacitive touch screen allows for easy navigation of the VWR® moisture analyzers menus. The IR proximity sensors allow for hands free operation, easily program the sensors to perform actions such as opening/closing drying chamber lid, zeroing, taring, or starting an experiment.
Store up to 5000 products, 200 drying modes, 5000 drying reports, and 100 users within the VWR® Moisture Analyzers databases.
Certifications: C-UL(US) and CE.
Delivery information: Includes moisture analyzer and aluminum pan.
Specifications for Product Family
Spécifications
- Catalog No:
- 76290-332
- 76290-334
- 76289-770
- 76289-772
- 76289-774
- 76289-740
- Calibration:
- External
- External
- External
- External
- External
- External
- Communication interface:
- RS232, USB (A and B), Wireless connection, Ethernet
- RS232, USB (A and B), Wireless connection, Ethernet
- RS232, USB (A and B), Wireless connection, Ethernet
- RS232, USB (A and B), Wireless connection, Ethernet
- RS232, USB (A and B), Wireless connection, Ethernet
- RS232, USB (A and B), Wireless connection, Ethernet
- Electrical:
- 120 V
- 120 V
- 120 V
- 120 V
- 120 V
- 120 V
- Operating temperature range:
- 10...40 °C
- 10...40 °C
- 10...40 °C
- 10...40 °C
- 10...40 °C
- 10...40 °C
- Plug type:
- Nema 5-15R
- Nema 5-15R
- Nema 5-15R
- Nema 5-15R
- Nema 5-15R
- Nema 5-15R
Spécifications
Product Family Options
Informations sur le produit
- DésignationModèleDimensionsPlateau de pesée/taille de la plateformeReadabilityRepeatabiltyWeighing CapacityAvailabilityPrice
- Halogen Heating ModuleVWR-54M.H33.8×20.6×19.7 cm9 cm0.1 mg/0.0001%0.05% (2 g sample)
0.01% (10 g sample)50 gDiscontinued-Spécifications:More
Specifications
Cat. No.76290-332CalibrationExternalCommunication interfaceRS232, USB (A and B), Wireless connection, EthernetElectrical120 VOperating temperature range10...40 °CPlug typeNema 5-15R - Halogen Heating ModuleVWR-53M.H33.8×20.6×19.7 cm9 cm1 mg/0.001%0.05% (2 g sample)
0.01% (10 g sample)50 gDiscontinued-Spécifications:More
Specifications
Cat. No.76290-334CalibrationExternalCommunication interfaceRS232, USB (A and B), Wireless connection, EthernetElectrical120 VOperating temperature range10...40 °CPlug typeNema 5-15R - Halogen Heating ModuleVWR-213M.H33.8×20.6×19.7 cm9 cm1 mg/0.001%0.05% (2 g sample)
0.01% (10 g sample)210 gDiscontinued-Spécifications:More
Specifications
Cat. No.76289-770CalibrationExternalCommunication interfaceRS232, USB (A and B), Wireless connection, EthernetElectrical120 VOperating temperature range10...40 °CPlug typeNema 5-15R - IR Emitter Heating ModuleVWR-54M.I33.8×20.6×19.7 cm9 cm0.1 mg/0.0001%0.05% (2 g sample)
0.01% (10 g sample)50 gLog in pour la disponibilitéEach$6,506.13Spécifications:More
Specifications
Cat. No.76289-772CalibrationExternalCommunication interfaceRS232, USB (A and B), Wireless connection, EthernetElectrical120 VOperating temperature range10...40 °CPlug typeNema 5-15R - IR Emitter Heating ModuleVWR-53M.I33.8×20.6×19.7 cm9 cm1 mg/0.001%0.05% (2 g sample)
0.01% (10 g sample)50 gLog in pour la disponibilitéEach$5,373.59Spécifications:More
Specifications
Cat. No.76289-774CalibrationExternalCommunication interfaceRS232, USB (A and B), Wireless connection, EthernetElectrical120 VOperating temperature range10...40 °CPlug typeNema 5-15R - IR Emitter Heating ModuleVWR-213M.I33.8×20.6×19.7 cm9 cm1 mg/0.001%0.05% (2 g sample)
0.01% (10 g sample)210 gLog in pour la disponibilitéEach$5,693.06Spécifications:More
Specifications
Cat. No.76289-740CalibrationExternalCommunication interfaceRS232, USB (A and B), Wireless connection, EthernetElectrical120 VOperating temperature range10...40 °CPlug typeNema 5-15R
Accessories
Informations sur le produit
- Catalog #AvailabilityPrice
- Weighing pans, aluminum, Ø 90 mm