Bone, cendre 20 Mesh
25 kg
  CA11020-386
 :  FA201-25KG
Bone, cendre 20 Mesh
  CA11020-386
 :  
Bone, cendre 20 Mesh
25 kg
 :  
$563.00
Each (25kg)
Ebonex bone ash is obtained by calcinating degelatinized bone up to a temperature of approximately 1100°C. Afterwards, the bone ash is cooled and milled (minimum of 95% through a 325 mesh size sieve).
Bone ash has a direct influence in improving the casting quality, because of the following intrinsical benefits:
  • Has excellent non-wetting properties
  • Is chemically inert and free of organic matters
  • Has very high heat transfer resistance
  • Increases the life expectancy of refractory and equipment
  • Taille:
    25 kg