Wildco® Plankton Tow Nets
Fournisseur: Science First
Certificats
À propos de cet article
Nets allow horizontal or vertical tows for plankton analysis.
- Collect plankton samples for analysis
The simplest method for collecting plankton samples. Usually towed by boats at sppeds of 1 to 3 knots. Tow Nets consist of a single hoop, bridle, net, and collection bucket.
Frequently Bought Together