MD-TMS™ Trace Mineral Supplement, American Type Culture Collection
Catalogue # MSPP-MDTMS
Fournisseur: AMERICAN TYPE CULTURE COLLECTION MS
Numéro CAS:
Spécifications
- Pack type:Vial
- Description:MD-TMS™ Trace Mineral Supplement
- Size:5×10 ml
- Cat. no.:MSPP-MDTMS
- Supplier No.:MD-TMS
Spécifications
À propos de cet article
ATCC® Trace mineral supplement is used as an additive in many ATCC® recommended media, especially for cultivating extremophiles. It speeds media preparation and reduces possible lab errors by combining several trace mineral ingredients into one sterile solution.
Packaging: Product is provided in an individual unit of use (10 ml).