ESD Safe Coil Cords, Botron
À propos de cet article
Coil cords are built for extended use and withstanding the labors of the work environment. It can be used in many applications from bench to personal grounding. Non-tangle and high retention coil make this a great choice for a hassle free path to ground.
- Lighter weight for better portability
- Reliable path to ground
- Available in lengths of 6' and 10' long
- Built in 1 meg Ohm ¹/₄ watt resistor at snap end
Cords are made of black PVC cord and 7 strand tinsel wire with superior strain reliefs at both ends.
Available with a spring loaded ¹/₈" (4 mm) socket connector to standard banana jack. Each cord comes with a slip on alligator clip that attaches to the banana jack for greater expansion of use.
Coil cords are ideal for personal work station grounding, wrist straps and smocks, as well as portable grounding with the included alligator clip. The non-slip banana jack provides an excellent connection to both common point and bench grounds.
Certifications: Meets S1.1 of one to five pounds breakaway force.
2 Options Available Below
- L'article nécessite un contrôle de la température pour le stockage et la livraison avec des frais supplémentaires. Il n'est pas éligible au retour pour des raisons de sécurité et de qualité. Tenez compte des exigences avant d'acheter.
- Return Policy
Product Details & Documents
Coil cords are built for extended use and withstanding the labors of the work environment. It can be used in many applications from bench to personal grounding. Non-tangle and high retention coil make this a great choice for a hassle free path to ground.
- Lighter weight for better portability
- Reliable path to ground
- Available in lengths of 6' and 10' long
- Built in 1 meg Ohm ¹/₄ watt resistor at snap end
Cords are made of black PVC cord and 7 strand tinsel wire with superior strain reliefs at both ends.
Available with a spring loaded ¹/₈" (4 mm) socket connector to standard banana jack. Each cord comes with a slip on alligator clip that attaches to the banana jack for greater expansion of use.
Coil cords are ideal for personal work station grounding, wrist straps and smocks, as well as portable grounding with the included alligator clip. The non-slip banana jack provides an excellent connection to both common point and bench grounds.
Certifications: Meets S1.1 of one to five pounds breakaway force.
Product Family Options
Informations sur le produit
- DésignationColorLongueurAvailabilityPrice
- Supplier Number: B2108L'article nécessite un contrôle de la température pour le stockage et la livraison avec des frais supplémentaires. Il n'est pas éligible au retour pour des raisons de sécurité et de qualité. Tenez compte des exigences avant d'acheter.
- Supplier Number: B2110L'article nécessite un contrôle de la température pour le stockage et la livraison avec des frais supplémentaires. Il n'est pas éligible au retour pour des raisons de sécurité et de qualité. Tenez compte des exigences avant d'acheter.