Ladies Safety Glasses, Pink
76210-824
- Frame Color:Pink
- Cat. no.:76210-824
- Lens Coating:Anti-fog, scratch-resistant
- Lens Tint:Clear
Stylish protective eyewear combining fashion, fit, and function.
- Molded-in nose bridge
- Soft, non-slip rubber temple tips
- Impact-resistant
- Polycarbonate lens
Most suitable for small facial contours and provides 99.9% UV protection.
: Meets ANSI Z87.1 standards.