Kimble Chase KimCote® Volumetric Flasks, Class A , DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
Certificates
About this item
Flasks feature a KimCote® plastic coating for added safety.
- Autoclavable KimCote® safety coating reduces the hazards of shattered glass and leakage of toxic or corrosive chemicals
- A single graduation ring is blasted on the neck
- Calibrated to contain
- A Standard Taper ground glass stopper is supplied
- Marking spots on all sizes
- Designed from ASTM Specification E288, Class A requirements
Manufactured from 33 expansions, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.
Frequently Bought Together