VWR® Dual Purpose Scoops
Fournisseur: VWR International
Certificats
À propos de cet article
VWR® dual purpose scoops are a unique and economical solution that combines the function of a scoop and a weighboat into one product.
- Made from anti-static polystyrene that is waterproof and reduces the chance of materials sticking to the scoop
- Ships and stores flat and when ready to use, is easy to assemble by inserting the locking tab into the hole
- Locking tab feature enables a scoop shape to help retain materials
- Coated to stand up to powders, gels, and semi-solids
- Broad tip design keeps the scoop stable while on a scale, and enables materials to be easily scooped and dispensed
The weighing process has been streamlined, allowing users to simply sample or retrieve material in the scoop and place it directly on the scale. This eliminates the need for a weighboat or weighing paper and saves valuable space in the lab. When finished, the scoop is disposable.
Frequently Bought Together