ContactUsLinkComponent
Interleukin-6 (Human) ELISA Kit, Cayman Chemical
Catalogue # 75816-834
Numéro CAS:  
undefined
Interleukin-6 (Human) ELISA Kit, Cayman Chemical
Catalogue # 75816-834
Supplier Number:  501030-96
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Spécifications
  • Primary antibody reactivity:
    Human
  • Description:
    Interleukin-6 (human) ELISA Kit
  • Cat. no.:
    75816-834
  • Contents:
    Kit contains: ELISA Strip Plate, Biotin Conjugate, ELISA Standard, Streptavidin-HRP, Immunoassay Buffer B Concentrate, Polysorbate 20, 96-Well Cover Sheet, Substrate Solution, Wash Buffer Concentrate, and Stop Solution
  • Supplier No.:
    501030-96
  • No. of Tests:
    96 wells
Spécifications
À propos de cet article
An immunometric assay for the measurement of IL-6.
  • Measure IL-6 from human plasma, serum, synovial fluid, and other sample matrices
  • Rapid assay
  • Get results in under 3 hours
Interleukin-6 (IL-6) is produced by multiple cell types, including monocytes, macrophages, and T cells and is involved in many defense mechanisms including the immune response, hematopoiesis, bone metabolism, and acute phase reactions.
IL-6 promotes the proliferation of activated B-cells, acts as a B-cell differentiation factor, and stimulates the secretion of immunoglobulins.
It also induces the differentiation of naïve T cells into TH17 effector cells.
Cayman’s IL-6 (human) ELISA Kit is an immunometric (sandwich) ELISA that permits IL-6 measurements within the range of 7.8 to 250 pg/ml, with an LLOQ of 7.8 pg/ml.