To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Spécifications
- Primary antibody reactivity:Human
- Description:Interleukin-6 (human) ELISA Kit
- Cat. no.:75816-834
- Contents:Kit contains: ELISA Strip Plate, Biotin Conjugate, ELISA Standard, Streptavidin-HRP, Immunoassay Buffer B Concentrate, Polysorbate 20, 96-Well Cover Sheet, Substrate Solution, Wash Buffer Concentrate, and Stop Solution
- Supplier No.:501030-96
- No. of Tests:96 wells
Spécifications
À propos de cet article
An immunometric assay for the measurement of IL-6.
- Measure IL-6 from human plasma, serum, synovial fluid, and other sample matrices
- Rapid assay
- Get results in under 3 hours
Interleukin-6 (IL-6) is produced by multiple cell types, including monocytes, macrophages, and T cells and is involved in many defense mechanisms including the immune response, hematopoiesis, bone metabolism, and acute phase reactions.
IL-6 promotes the proliferation of activated B-cells, acts as a B-cell differentiation factor, and stimulates the secretion of immunoglobulins.
It also induces the differentiation of naïve T cells into TH17 effector cells.
Cayman’s IL-6 (human) ELISA Kit is an immunometric (sandwich) ELISA that permits IL-6 measurements within the range of 7.8 to 250 pg/ml, with an LLOQ of 7.8 pg/ml.