ContactUsLinkComponent
Magid® Gemstone Garnet Y60 Series Protective Spectacle, Magid Glove
Catalogue # 62300-104
Fournisseur:  Magid Glove
Small Business Enterprise
Magid® Gemstone Garnet Y60 Series Protective Spectacle, Magid Glove
Catalogue # 62300-104
Fournisseur:  Magid Glove
Supplier Number:  Y60BKC
Small Business Enterprise
Spécifications
  • Frame Color:
    Black
  • Cat. no.:
    62300-104
  • Lens Coating:
    Scratch-resistant/hardcoat
  • Lens Tint:
    Clear
  • Supplier No.:
    Y60BKC
  • Frame Material:
    Plastic
Spécifications
À propos de cet article
Magid® Gemstone Garnet Y60 series is a frameless style spectacle that features contoured lens for added front, side, brow and cheek protection.
  • Anti-scratch, clear lens
  • Black temples and an anti-scratch lens
This high-quality spectacle provides 99.9% protection against harmful ultraviolet light.
Certifications: Meets ANSI Z87.1-2010 high impact standards.