ContactUsLinkComponent
Nuclear Extraction Kit, Cayman Chemical Company
Nuclear Extraction Kit, Cayman Chemical Company
Catalog # 75816-726
CAS Number:  
undefined
Nuclear Extraction Kit, Cayman Chemical Company
Catalog # 75816-726
Supplier Number:  10009277-1
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    Kit contains: Hypotonic Buffer, Dithiothreitol, Inhibitor Cocktail, PBS, Phosphatase Inhibitors, Extraction Buffer, Reagent
  • Cat. no.:
    75816-726
  • Supplier No.:
    10009277-1
Specifications
About this item
For the quick and simple isolation of nuclear and cytoplasmic fractions from cultured cells and tissue homogenates
  • Isolate nuclear and cytoplasmic fractions from cell lysates and tissue homogenates
  • Resulting isolates can be used in Transcription Factor Assay Kits
  • Isolates can also be used for EMSA or western blotting
  • Each kit provides reagents for 50 extractions from approximately 10 million cells
Preparation of nuclear extracts is the first step in examining transcription factor activity. Cayman’s Nuclear Extraction is formulated for the quick and simple isolation of nuclear and cytoplasmic fractions from cultured cells and tissue homogenates that can be used successfully in Cayman’s Transcription Factor Assay kits. The proteins isolated using this kit can also be used in electrophoretic mobility shift assays (EMSA) and western blotting applications.