Sharp-Point Surgical Scissors
Sharp-Point Surgical Scissors, 140 mm (5.5"), Stainless steel, Sharp/Sharp
Catalog # 470105-650
Supplier: Pinetree Instruments
Fournisseur #: 22272
Jump to:
Product Details & Documents
Start Your Cuts Easily.
- Dissect without a scalpel
- Handles rounded for ambidextrous use
Sharp-pointed, straight surgical scissors allow the user to insert blades into tissue to start the cut. Larger scissors provide greater leverage for tough cutting; shorter scissors for finer work. Stainless steel.
Spécifications
- Désignation:Sharp-Point Surgical Scissors
- Longueur:140 mm (5.5")
- Matière:Stainless steel
- Blade Type:Sharp/Sharp